Anti-SEC63 Rabbit Polyclonal Antibody

Supplier: Proteintech
13978-1-AP-150UL
10094-270EA 402 USD
10094-270
Anti-SEC63 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:1000-1:10000

Type: Primary
Antigen: SEC63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR