Anti-HSPA1L Rabbit Polyclonal Antibody

Supplier: Proteintech
13970-1-AP-150UL
10088-514EA 402 USD
10088-514
Anti-HSPA1L Rabbit Polyclonal Antibody
Antibodies
HSPA1L (Hsp70-like protein 1, also known as HSP70-hom) is a member of Hsp70 family of heat shock proteins. Recently it has been reported that HSPA1L can interact with TLR4, activate dendritic cells and induce Th1 Response. This antibody well recognized the endogenous HSPA1L protein in multiple cell lines. (21730052)

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human endometrial cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: HSPA1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR