Anti-WARS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13944-1-AP-150UL
10097-064EA 402 USD
10097-064
Anti-WARS2 Rabbit Polyclonal Antibody
Antibodies
WARS2(tryptophan--tRNA ligase, mitochondrial) is also called mitochondrial TRPRS and belongs to the class-I aminoacyl-tRNA synthetase family. The gene encodes the protein containing 360-amino acid ,which has an N-terminal mitochondrial targeting signal that is cleaved after residue 18. WARS2 has characteristics of a class I aminoacyl-tRNA synthetase, including a classical Rossmann fold. It has 2 isoforms produced by alternative splicing.

Western Blot: Human Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: WARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR