Anti-FBXL14 Rabbit Polyclonal Antibody

Supplier: Proteintech
13934-1-AP-150UL
10086-900EA 402 USD
10086-900
Anti-FBXL14 Rabbit Polyclonal Antibody
Antibodies
FBXL14(F-box/LRR-repeat protein 14) is also named as FBL14 and belongs to the the F-box protein family,which are characterized by an approximately 40-amino acid F-box motif. It is part of a SCF (SKP1-cullin-F-box) ubiquitin-protein ligase complex. SNAIL1 is an unstable transcription factor that is phosphorylated by GSK3-beta in the nucleus, which triggers its nuclear export and subsequent ubiquitination and proteasomal degradation. FBXL14 is directed ubiquitination and degradation of SNAIL1 in human and mouse cell lines.

Western Blot: HepG2 Cells, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: FBXL14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR