Anti-PTPRE Rabbit Polyclonal Antibody

Supplier: Proteintech
13922-1-AP-150UL
10093-076EA 402 USD
10093-076
Anti-PTPRE Rabbit Polyclonal Antibody
Antibodies
Western Blot: HL-60 Cells, 1:200-1:1000

Type: Primary
Antigen: PTPRE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR