Anti-NKX2-5 Rabbit Polyclonal Antibody

Supplier: Proteintech
13921-1-AP-150UL
10091-136EA 402 USD
10091-136
Anti-NKX2-5 Rabbit Polyclonal Antibody
Antibodies
Homeobox-containing genes play critical roles in regulating tissue-specific gene expression essential for tissue differentiation, as well as determining the temporal and spatial patterns of development . NKX2-5 encoded by homeobox-containing genes that implicated in commitment to and/or differentiation of the myocardial lineage, and acts as a transcriptional activator of ANF in cooperation with GATA4. During embryonic development, NKX2-5 plays pivotal roles in specifying cardiac progenitors, cardiac morphogenesis, cardiomyocyte differentiation and conduction system development .

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: NKX2-5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR