Anti-BAG4 Rabbit Polyclonal Antibody

Supplier: Proteintech
13913-1-AP-150UL
10094-998EA 524.48 USD
10094-998
Anti-BAG4 Rabbit Polyclonal Antibody
Antibodies
BAG4, also known as SODD (the silencer of death domain), belongs to the BAG family of anti-apoptotic proteins. BAG4 binds to the tumor necrosis factor receptor 1 (TNFR1) and block signaling for subsequent activation of apoptosis. This antibody detects 66-70 kDa full-length BAG4 protein and 45 kDa, 60 kDa C-terminal fragments, which was also observed by Hedda et al. (12706861).

Western Blot:K-562 Cells, 1:500-1:5000

Type: Primary
Antigen: BAG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR