Anti-HPS5 Rabbit Polyclonal Antibody

Supplier: Proteintech
13901-1-AP-150UL
10088-440EA 402 USD
10088-440
Anti-HPS5 Rabbit Polyclonal Antibody
Antibodies
Hermansky-Pudlak syndrome 5 protein (HPS-5), also named Ruby-eye protien 2 homolog (Ru2), plays a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. This protein interacts with Hermansky-Pudlak syndrome 6 protein and may interact with the cytoplasmic domain of integrin, alpha-3. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 5.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: HPS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR