Anti-D2HGDH Rabbit Polyclonal Antibody

Supplier: Proteintech
13895-1-AP-150UL
10085-638EA 402 USD
10085-638
Anti-D2HGDH Rabbit Polyclonal Antibody
Antibodies
D2HGDH(D-2-hydroxyglutarate dehydrogenase, mitochondrial) is also named as D2HGD and belongs to the FAD-binding oxidoreductase/transferase type 4 family.It catalyzes the oxidation of D-2-hydroxyglutarate to alpha-ketoglutarate.Defects in D2HGDH are the cause of D-2-hydroxyglutaric aciduria type 1 (D2HGA1).It has 2 isoforms produced by alternative splicing.

Western Blot: Human Liver Tissue, 1:200-1:2000; IHC: Human Liver cancer Tissue, 1:20-1:200; IP: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: D2HGDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR