Anti-NDUFAF2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13891-1-AP-150UL
10090-956EA 402 USD
10090-956
Anti-NDUFAF2 Rabbit Polyclonal Antibody
Antibodies
Mimitin, also named as MMTN, B17.2L, NDUFAF2 and NDUFA12L, is a small mitochondrial protein upregulated by proinflammatory cytokines at the transcriptional and protein levels. It is reported as a possible chaperone for assembly of mitochondrial complex I. The MW of Mimitin is 17-25 kDa.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: NDUFAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR