Anti-SIAH1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13886-1-AP-150UL
10094-638EA 402 USD
10094-638
Anti-SIAH1 Rabbit Polyclonal Antibody
Antibodies
SIAH1, also named as HUMSIAH, Siah-1a and Siah-1S(21kd), belongs to the SINA (Seven in absentia) family. SIAH1 is known to cause indirect degradation of beta-catenin through formation of a complex with Siah-interacting protein (SIP), Skp1 and Ebi. SIAH1 can form heterodimer or homodimer such as Siah-1*Siah-1(70 or 62kd), Siah-1*Siah-1S(42kd) and Siah-1S*Siah-1S(51-56kd). Importantly, results from in vitro soft agar assay demonstrated that Siah-1S displays a promotion effect on cells tumorigenicity.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: SIAH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR