Anti-DGKH Rabbit Polyclonal Antibody

Supplier: Proteintech
13873-1-AP-150UL
10085-796EA 402 USD
10085-796
Anti-DGKH Rabbit Polyclonal Antibody
Antibodies
DGKH(Diacylglycerol kinase eta) is also named as DGK-eta and belongs to the eukaryotic diacylglycerol kinase family. It is a key protein in the lithium-sensitive phosphatidyl inositol pathway, and more recently a link between environmental stress and schizophrenia and possibly bipolar disorder has been revealed. DGKH has 3 isoforms produced by alternative splicing.Two isoforms DGKH1 and DGKH2 with the molecular weight of 128 kDa and 135 kDa shows diacylglycerol kinase activity; however, the 2 isoforms differed in several other characteristics.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: DGKH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR