Anti-VPS41 Rabbit Polyclonal Antibody

Supplier: Proteintech
13869-1-AP-150UL
10097-026EA 402 USD
10097-026
Anti-VPS41 Rabbit Polyclonal Antibody
Antibodies
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. VPS41 (vacuolar protein sorting 41) is the human ortholog of yeast Vps41 protein. Yeast Vps41 is a subunit of AP3 (heterotetrameric adaptor protein complex 3) and HOPS (homotypic fusion and protein transport) complexes and is involved in trafficking of proteins from the late Golgi to the vacuole. In mammalian cells, Vps41 was found to be a member of HOPS complex, mediating the trafficking of proteins to lysosomes . It has been reported that human VPS41 has a HOPS-independent function in the LAMP carrier pathway and also participates in lysosomal maturation, probably as part of the HOPS complex .

Western Blot: HepG2 Cells, 1:200-1:1000

Type: Primary
Antigen: VPS41
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR