Anti-TIMM44 Rabbit Polyclonal Antibody

Supplier: Proteintech
13859-1-AP-150UL
10096-004EA 402 USD
10096-004
Anti-TIMM44 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:2000; IF:A549 Cells, 1:20-1:200

Type: Primary
Antigen: TIMM44
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR