Anti-SORBS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13854-1-AP-150UL
10095-002EA 402 USD
10095-002
Anti-SORBS1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:3T3-L1 Cells, 1:200-1:2000

Type: Primary
Antigen: SORBS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR