Anti-JAKMIP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13846-1-AP-150UL
10089-068EA 402 USD
10089-068
Anti-JAKMIP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse spleen Tissue, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200; IP:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: JAKMIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR