Anti-ARTS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13821-1-AP-150UL
10083-174EA 402 USD
10083-174
Anti-ARTS1 Rabbit Polyclonal Antibody
Antibodies
ARTS1 is a 100 kDa, monomeric, soluble zinc aminopeptidase that belongs to the M1 family of metallo-peptidases. ERAP1 is induced by interferon-gama and can degrade peptides 9-15 residues long.It can be glycosylated to 120KDa and form homodimers and heterodimers.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: ARTS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR