Anti-NISCH Rabbit Polyclonal Antibody

Supplier: Proteintech
13813-1-AP-150UL
10091-118EA 402 USD
10091-118
Anti-NISCH Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: NISCH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR