Anti-NPHP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
13812-1-AP-150UL
10091-250EA 537.7 USD
10091-250
Anti-NPHP4 Rabbit Polyclonal Antibody
Antibodies
NPHP4, also named as KIAA0673, is a cilia-associated protein which involved in the organization of apical junctions in kidney cells together with NPHP1 and RPGRIP1L/NPHP8. It is necessary for normal photoreceptor ribbon synapse maintenance and outer segment formation, and for sperm development.

IHC: Human Heart Tissue, 1:20-1:200; Western Blot: Mouse Brain Tissue, 1:200-1:2000; IF: HTERT-RPE1 Cells, 1:10-1:100

Type: Primary
Antigen: NPHP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR