Anti-CDH3 Rabbit Polyclonal Antibody

Supplier: Proteintech
13773-1-AP-150UL
10091-902EA 402 USD
10091-902
Anti-CDH3 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF:A431 Cells, 1:20-1:200

Type: Primary
Antigen: CDH3
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR