Anti-GPRIN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13771-1-AP-150UL
10087-806EA 402 USD
10087-806
Anti-GPRIN1 Rabbit Polyclonal Antibody
Antibodies
GPRIN1 (G protein-regulated inducer of neurite outgrowth 1), also named GRIN1, may be involved in neurite outgrowth. Western blotting analysis has confirmed its expression in mouse neuroblastoma and rat pheochromocytoma cells. And both GPRIN1 and GNAO1 are membrane-bound proteins that are enriched in the growth cones of neurites . It has 2 isoforms produced by alternative splicing. This antibody detects GRIN1 at bands around 110 kDa.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IP: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: GPRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR