Anti-ADAP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13706-1-AP-150UL
10082-500EA 402 USD
10082-500
Anti-ADAP2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Placenta Tissue, 1:200-1:1000; IHC: Human spleen, 1:20-1:200

Type: Primary
Antigen: ADAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR