Anti-WDR19 Rabbit Polyclonal Antibody

Supplier: Proteintech
13647-1-AP-150UL
10097-094EA 402 USD
10097-094
Anti-WDR19 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:200-1:2000; IF: HTERT-RPE1 Cells, 1:20-1:200; IHC: Human prostate hyperplasia Tissue, 1:20-1:200

Type: Primary
Antigen: WDR19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR