Anti-UIMC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13642-1-AP-150UL
10096-768EA 402 USD
10096-768
Anti-UIMC1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Colon Tissue, 1:200-1:1000; IF: Hela Cells, 1:10-1:100; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: UIMC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR