Anti-TROAP Rabbit Polyclonal Antibody

Supplier: Proteintech
13634-1-AP-150UL
10096-394EA 402 USD
10096-394
Anti-TROAP Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:500-1:5000; IF: MCF-7, 1:10-1:100

Type: Primary
Antigen: TROAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR