Anti-CAMK1D Rabbit Polyclonal Antibody

Supplier: Proteintech
13613-1-AP-150UL
10084-024EA 402 USD
10084-024
Anti-CAMK1D Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Thymus Tissue, 1:1000-1:10000

Type: Primary
Antigen: CAMK1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR