Anti-FBXO22 Rabbit Polyclonal Antibody

Supplier: Proteintech
13606-1-AP-150UL
10086-908EA 402 USD
10086-908
Anti-FBXO22 Rabbit Polyclonal Antibody
Antibodies
FBXO22, also named as FBX22, is a substrate-recognition of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. This antibody recognizes isoform 1 (45 kDa) and isoform 3 (30-33 kDa).

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human tonsil Tissue, 1:20-1:200

Type: Primary
Antigen: FBXO22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR