Anti-IL2RB Rabbit Polyclonal Antibody

Supplier: Proteintech
13602-1-AP-150UL
10088-830EA 402 USD
10088-830
Anti-IL2RB Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:200-1:1000

Type: Primary
Antigen: IL2RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR