Anti-PRDX6 Rabbit Polyclonal Antibody

Supplier: Proteintech
13585-1-AP-150UL
10092-762EA 524.48 USD
10092-762
Anti-PRDX6 Rabbit Polyclonal Antibody
Antibodies
Prdx6(Peroxiredoxin-6) , a unique member of the peroxiredoxin family of antioxidants, is highly expressed in liver and protects cells from oxidative damage by reducing H2O2 and various lipid Peroxides.It is also named as AOP2, KIAA0106. It can form a dimer.PRDX6 is expressed in all major organs, with a particularly high level in lung.

Western Blot:A549 Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200; IF: Hela Cells, 1:20-1:200

Type: Primary
Antigen: PRDX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR