Anti-ABCG1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13578-1-AP-150UL
10082-354EA 537.7 USD
10082-354
Anti-ABCG1 Rabbit Polyclonal Antibody
Antibodies
IHC: Human cervical cancer Tissue, 1:20-1:200; Western Blot: Multi-Cells/Tissue, 1:500-1:5000

Type: Primary
Antigen: ABCG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR