Anti-MNDA Rabbit Polyclonal Antibody

Supplier: Proteintech
13576-1-AP-150UL
10090-326EA 402 USD
10090-326
Anti-MNDA Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:1000-1:10000

Type: Primary
Antigen: MNDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR