Anti-ARFGAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13571-1-AP-150UL
10083-050EA 402 USD
10083-050
Anti-ARFGAP1 Rabbit Polyclonal Antibody
Antibodies
Protein coats deform flat lipid membranes into buds and capture membrane proteins to form transport vesicles. The assembly-disassembly cycle of the COPI coat on Golgi membranes is coupled to the GTP-GDP cycle of the small G protein ARF1. ARFGAP1 contributes to vesicle budding by increasing the GTPase activity of ARF1, because it promotes hydrolysis of the ARF1-bound GTP and thus, is required for the dissociation of coat proteins from Golgi-derived membranes and vesicles, a prerequisite for vesicle's fusion with target compartment. for ARFGAP1 may has a role in regulating the GTPase activity and neuronal toxicity of LRRK2; reciprocally, LRRK2 phosphorylates ArfGAP1 and is required for ArfGAP1 neuronal toxicity.

Western Blot:PC-3 Cells, 1:500-1:5000; IHC: Human prostate cancer, 1:20-1:200

Type: Primary
Antigen: ARFGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR