Anti-CYP2J2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13562-1-AP-150UL
10085-526EA 402 USD
10085-526
Anti-CYP2J2 Rabbit Polyclonal Antibody
Antibodies
CYP2J2(Cytochrome P450 2J2) belongs to the cytochrome P450 family. This enzyme metabolizes arachidonic acid predominantly via a NADPH-dependent olefin epoxidation to all four regioisomeric cis-epoxyeicosatrienoic acids. The human CYP2J member, CYP2J2, is shown to be expressed in several tissues such as the small intestine, liver, heart, kidney, and lung and may play a role in the oxidative bioactivation of arachidonic acid.

Western Blot: Mouse Skeletal Muscle Tissue, 1:200-1:2000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CYP2J2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR