Anti-COG7 Rabbit Polyclonal Antibody

Supplier: Proteintech
13552-1-AP-150UL
10084-974EA 402 USD
10084-974
Anti-COG7 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Y79 Cells, 1:500-1:5000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: COG7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR