Anti-PDSS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13544-1-AP-150UL
10092-058EA 524.48 USD
10092-058
Anti-PDSS2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SGC-7901 Cells, 1:200-1:2000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: PDSS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR