Anti-CROT Rabbit Polyclonal Antibody

Supplier: Proteintech
13543-1-AP-150UL
10085-208EA 402 USD
10085-208
Anti-CROT Rabbit Polyclonal Antibody
Antibodies
CROT, also named as COT, belongs to the carnitine/choline acetyltransferase family. It is a beta-oxidation of fatty acids. The highest activity concerns the C6 to C10 chain length substrate. CROT converts the end product of pristanic acid beta oxidation, 4,8-dimethylnonanoyl-CoA, to its corresponding carnitine ester. Carnitine palmitoyltransferase (CPT) deficiencies are common disorders of mitochondrial fatty acid oxidation. The CPT system is made up of two separate proteins located in the outer (CPT1) and inner (CPT2) mitochondrial membranes. CROT is an active forms for carnitine octanoyltransferase. This antibody can bind the close sequences genes.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: CROT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR