Anti-PAK6 Rabbit Polyclonal Antibody

Supplier: Proteintech
13539-1-AP-150UL
10091-764EA 402 USD
10091-764
Anti-PAK6 Rabbit Polyclonal Antibody
Antibodies
PAK6(p21-activated kinase 6) belongs to the PAK family.This protein plays a role in the regulation of gene transcription and may protect cells from apoptosis through phosphorylation of BAD.Northern blot analysis revealed that PAK6 is highly expressed in testis and prostate tissues.

Western Blot:PC-3 Cells, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PAK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR