Anti-MARVELD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13515-1-AP-150UL
10089-994EA 402 USD
10089-994
Anti-MARVELD2 Rabbit Polyclonal Antibody
Antibodies
MARVELD2, also named as Tricellulin or TRIC, is a 558 amino acid protein, which contains one MARVEL domain. MARVELD2 localizes the cell membrane and plays a role in the formation of the epithelial barriers. MARVELD2, is a first integral membrane protein that is concentrated at the vertically oriented tight junction strands of tricellular contacts. MARVELD2 may be a component to maintain the integrity for peripheral nervous system myelin function and morphology.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000

Type: Primary
Antigen: MARVELD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR