Anti-p115, USO1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13509-1-AP-150UL
10090-922EA 524.48 USD
10090-922
Anti-p115, USO1 Rabbit Polyclonal Antibody
Antibodies
p115, also known as USO1, TAP (transcytosis-associated protein) or VDP (vesicle docking protein) is a general vesicular transport factor and plays an important role at different steps of vesicular transport. It is a 962-residue peripheral membrane protein which recycles between the cytosol and the Golgi apparatus during interphase . p115 forms stable homodimers . Rab1 recruits p115 to coat protein complex II (COPII) vesicles during budding from the endoplasmic reticulum, where p115 interacts directly with a select set of SNARE proteins . p115 is required for intra-Golgi transport, and also functions in endoplasmic reticulum to Golgi trafficking, Golgi biogenesis and exocytotic transport .

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200; IF: Hela Cells, 1:20-1:200; IP: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: p115, USO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR