Anti-GPX7 Rabbit Polyclonal Antibody

Supplier: Proteintech
13501-1-AP-150UL
10087-824EA 402 USD
10087-824
Anti-GPX7 Rabbit Polyclonal Antibody
Antibodies
GPX7(Glutathione peroxidase 7) is also named as GPX6,CL683,GSHPx-7 and belongs to the glutathione peroxidase family.It shares approximately 45% amino acid sequence with other GPX family members.Because GPX7 expression protected cells from acidic bile acidinduced DNA damage, it may protect oesophageal epithelial cells from acidic bile acid-induced apoptosis.

Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: GPX7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR