Anti-SPINK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13477-1-AP-150UL
10095-098EA 402 USD
10095-098
Anti-SPINK1 Rabbit Polyclonal Antibody
Antibodies
IHC: Human pancreas, 1:10-1:100

Type: Primary
Antigen: SPINK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR