Anti-SDCCAG8 Rabbit Polyclonal Antibody

Supplier: Proteintech
13471-1-AP-150UL
10093-754EA 402 USD
10093-754
Anti-SDCCAG8 Rabbit Polyclonal Antibody
Antibodies
SDCCAG8, also named as CCCAP and NY-CO-8, plays a role in the establishment of cell polarity and epithelial lumen formation. It may play a role in ciliogenesis. Loss of SDCCAG8 function as a cause of a retinal-renal ciliopathy and validates exome capture analysis for broadly heterogeneous single-gene disorders. SDCCAG8 is localized at both centrioles and interacts directly with OFD1 (oral-facial-digital syndrome 1), which is associated with NPHP-RC.

Western Blot:PC-3 Cells, 1:200-1:2000; IF: Hela Cells, 1:20-1:200

Type: Primary
Antigen: SDCCAG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR