Anti-CETP Rabbit Polyclonal Antibody

Supplier: Proteintech
13459-1-AP-150UL
10084-682EA 524.48 USD
10084-682
Anti-CETP Rabbit Polyclonal Antibody
Antibodies
Cholesteryl ester transfer protein (CETP) is a plasma glycoprotein which transfers cholesteryl esters, triglycerides, and phospholipids between lipoproteins. In plasma, CETP is mainly associated with HDL particles and catalyzes the transfer of cholesteryl esters from HDL to apolipoprotein (apo) B-containing plasma lipoproteins. Low plasma concentrations of high-density lipoprotein-cholesterol (HDL-C) have long been recognized as a major, independent cardiovascular disease (CVD) risk factor. It demonstrated that altered level of plasma CETP was associated with the increased risk of CVD. (17991755, 23477743)

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: CETP
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR