Anti-GIMAP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13445-1-AP-150UL
10087-544EA 402 USD
10087-544
Anti-GIMAP2 Rabbit Polyclonal Antibody
Antibodies
This protein was not eliminated clearly. It belongs to GTPase of the immunity-associated protein family. One of its homologous protein, GIMAP1, runs as a 30kd protein in SDS-PAGE, lower than the calculated molecular weight. This antibody detected one 28kd band in several tissues.

Western Blot: Human Heart Tissue, 1:200-1:2000

Type: Primary
Antigen: GIMAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR