Anti-SFRS17A Rabbit Polyclonal Antibody

Supplier: Proteintech
13441-1-AP-150UL
10094-456EA 402 USD
10094-456
Anti-SFRS17A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: HT-1080 Cells, 1:10-1:100

Type: Primary
Antigen: SFRS17A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR