Anti-CYP51A1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13431-1-AP-150UL
10085-546EA 402 USD
10085-546
Anti-CYP51A1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:PC-3 Cells, 1:500-1:5000

Type: Primary
Antigen: CYP51A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR