Anti-NDUFV3 Rabbit Polyclonal Antibody

Supplier: Proteintech
13430-1-AP-150UL
10090-994EA 402 USD
10090-994
Anti-NDUFV3 Rabbit Polyclonal Antibody
Antibodies
NDUFV3(NADH dehydrogenase [ubiquinone] flavoprotein 3, mitochondrial) is also named as CI-9kD,Renal carcinoma antigen NY-REN-4 and belongs to the complex I NDUFV3 subunit family.It is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: NDUFV3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR