Anti-GSK3A Rabbit Polyclonal Antibody

Supplier: Proteintech
13419-1-AP-150UL
10087-900EA 402 USD
10087-900
Anti-GSK3A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human serum Tissue, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: GSK3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR