Anti-KIF22 Rabbit Polyclonal Antibody

Supplier: Proteintech
13403-1-AP-150UL
10089-232EA 402 USD
10089-232
Anti-KIF22 Rabbit Polyclonal Antibody
Antibodies
The forces required for microtubule-based movements are generated by microtubule-associated motor proteins that include cytoplasmic dynein and kinesin.KIF22 also known as Kid is a kinesin-like DNA-binding protein known to be involved in chromosome movement during mitosis.Recent data suggest that Kid-mediated anaphase/telophase chromosome compaction prevents formation of multinucleated cells, which is especially important during the very early stages of development.

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: KIF22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR