Anti-MIP-3-beta Rabbit Polyclonal Antibody

Supplier: Proteintech
13397-1-AP-150UL
10090-234EA 402 USD
10090-234
Anti-MIP-3-beta Rabbit Polyclonal Antibody
Antibodies
CCL19, also known as MIP-3β, is one of several CC cytokine genes clustered on the p-arm of chromosome 9. CCL19 may play a role in normal lymphocyte recirculation and homing. It also plays an important role in trafficking of T cells in thymus, and in T cell and B cell migration to secondary lymphoid organs. It specifically binds to chemokine receptor CCR7.

Western Blot:A549 Cells, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: MIP-3-beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR